Would you like to log in again?
Anti Beta2 Glycoprotein1 IgG ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM110
103389-112EA 516.54 USD InStock
103389-112
Anti Beta2 Glycoprotein1 IgG ELISA, Eagle Biosciences
Assays ELISAs
AntiBeta 2 Glycoprotein 1 IgG ELISA measures IgG class Ab against Beta 2-Glycoprotein 1 in samples.
  • Dynamic Range: 10 - 160AU/mL
  • Sensitivity: 0.47 AU/mL
  • Incubation: 2 hours
The antiphospholipid syndrome (APS) is a disorder that  presents peculiar symptoms: arterial and venous thrombosis, thrombocytopenia, ulcers of the lower limbs, hemolytic anemia, loss of the fetus during pregnancy and is associated with the presence of antiphospholipid antibodies. Antiphospholipid antibodies represent a large and heterogeneous immunoglobulins group, including anticardiolipin antibodies and lupus anticoagulant. The former are diagnosed by on their reactivity with cardiolipin or other anionic phospholipids in ELISA test, while the latter is detected in phospholipid-dependent coagulation tests (aPTT, KCT, dRVVT).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty