Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti Beta2 Glycoprotein 1 IgM ELISA, Eagle Biosciences

Supplier: Eagle Biosciences
DCM111
103389-114EA 516.54 USD InStock
103389-114
Anti Beta2 Glycoprotein 1 IgM ELISA, Eagle Biosciences
Assays ELISAs
AntiBeta 2 Glycoprotein 1 IgM ELISA measures IgM class Ab against Beta 2-Glycoprotein 1 in samples.

  • Dynamic Range: 10 - 160AU/mL
  • Sensitivity: 0.11 AU/mL
  • Incubation: 2 hours

The antiphospholipid syndrome (APS) is a disorder that  presents peculiar symptoms: arterial and venous thrombosis, thrombocytopenia, ulcers of the lower limbs, hemolytic anemia, loss of the fetus during pregnancy and is associated with the presence of antiphospholipid antibodies. Antiphospholipid antibodies represent a large and heterogeneous immunoglobulins group, including anticardiolipin antibodies and lupus anticoagulant. The former are diagnosed by on their reactivity with cardiolipin or other anionic phospholipids in ELISA test, while the latter is detected in phospholipid-dependent coagulation tests (aPTT, KCT, dRVVT). In early '90s it was observed that antiphospholipid antibodies are not directed against anionic phospholipids, as long it was considered, but they react with plasma proteins bound to anionic (phospholipidic) surfaces. In fact about cardiolipin, it was observed the need of a cofactor for antibodies binding, this cofactor was identified in β2 glycoprotein 1 (β2-GP1). The β2 glycoprotein 1 is a plasma glycoprotein of molecular weight of 50 kD, which is complexed to lipoproteins for 40%. A patient with clinical suspicion of APS, who has an high titre of anti-beta2GP1 antibodies, even if  LAC or anti-cardiolipin antibodies are negative,  has a strong chance of having the antiphospholipid syndrome, because these antibodies recognize β2-GP1 bound to the surface of phospholipid (cardiolipin). These proteins seem to express their Antigensicity only after contact with specific areas such as the anionic phospholipid surface or very hydrophilic plastic surfaces. The importance of ELISA anti-phospholipids test, including  the anti-β2-GP1 test, lies in the fact that a positive test if associated with one or more of the symptoms can confirm a diagnosis of APS.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty