Would you like to log in again?
Anti Cardiolipin IgM ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM112
103389-116EA 555.77 USD InStock
103389-116
Anti Cardiolipin IgM ELISA, Eagle Biosciences
Assays ELISAs
Anti Cardiolipin IgM ELISA measures IgM class auto-Ab in serum or plasma.
  • Dynamic Range: 5 - 80 AU/ml
  • Sensitivity: 0.12 AU/ml
  • Incubation: 2 hours
The first study on the anti-phospholipid antibodies began in 1906 when Wasserman introduced a serological test for syphilis. In 1942 it was discovered that the active component is a phospholipid called Cardiolipina. In the 1950's it was observed that a large number of people appeared to be positive for syphilis tests but did not show any evidence of disease. Initially the phenomenon was classified as a series of false positive syphilis tests, before a more accurate analysis revealed, for this group of patients, a high prevalence of autoimmune disorders including SLE and Sjögrens syndrome.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty