Would you like to log in again?
Anti Cardiolipin IgG ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM113
103389-118EA 555.77 USD InStock
103389-118
Anti Cardiolipin IgG ELISA, Eagle Biosciences
Assays ELISAs
Anti Cardiolipin IgG ELISA measures IgG class auto-Ab in serum or plasma.
  • Dynamic Range: 16 - 64 µgEq/m
  • Sensitivity: 0.08 µgEq/m
  • Incubation: 2 hours
The first study on the anti-phospholipid antibodies began in 1906 when Wasserman introduced a serological test for syphilis. In 1942 it was discovered that the active component is a phospholipid called Cardiolipina. In the 1950's it was observed that a large number of people appeared to be positive for syphilis tests but did not show any evidence of disease. Initially the phenomenon was classified as a series of false positive syphilis tests, before a more accurate analysis revealed, for this group of patients, a high prevalence of autoimmune disorders including SLE and Sjögrens  syndrome.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty