Would you like to log in again?
Mouse / Rat Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ADU39-K010
103389-146EA 1222.69 USD InStock
103389-146
Mouse / Rat Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences
Assays ELISAs
Adrenaline Sensitive ELISA is intended to detect Adrenaline in biological fluids by a microplate .
  • Dynamic Range: 0.15 - 25 ng/ml
  • Sensitivity: 1.6 pg/mL
  • Incubation: overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty