Would you like to log in again?
HAHA (Human Anti-Human Antibody) ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
HAH31-K01
103389-156EA 1176.92 USD InStock
103389-156
HAHA (Human Anti-Human Antibody) ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Human HAHA ELISA determines human anti-human IgG antibody in serum or plasma samples.
  • Dynamic Range: 0 - 27 ug/mL
  • Sensitivity: 0.1 ug/mL
  • Incubation: 2 hours
Clinically, humanized monoclonal antibodies (IgG) and their fragments are used in vivo diagnosis procedure (radionuclides) and treatment for patients with various diseases. In patients, even a single dose injection of a humanized monoclonal antibody or its fragment may induce immune response directed against this foreign protein (immunogen). Also, people with autoimmune diseases, such as rheumatoid arthritis, lupus, etc. produce autoantibody against human IgG. In the circulation, the presence of human antibody against human IgG would bind to the injected humanized antibody therapeutics or diagnosis and, therefore, diminish the efficacy of either in-vivo diagnosis or treatment. Especially, the HAHA would increase the risk of anaphylactic complications to subsequent administration of the humanized monoclonal antibody-based therapy. The presence of HAHA in patient serum or plasma specimens may cause both false positive and false negative immunoassay test results depending on assay principles and monoclonal antibodies used in the assay system. This Eagle Biosciences HAHA ELISA Assay Kit is a ready-to-use test kit with a breakable well microtiter plate and simple test procedures. It also provides a wide measurement range without high dose “hook” effect.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty