Would you like to log in again?
Fecal C. Difficile Toxin B Qualitative ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
DFB35-K01
103389-164EA 568.85 USD InStock
103389-164
Fecal C. Difficile Toxin B Qualitative ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Clostridium Difficile Toxin B ELISA is intended for qualitative detection of C
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2 hours
Clostridium difficile is a common pathogen and a major cause of infectious diarrhea in hospitalized patients. While most strains produce both Toxin A and Toxin B, some only produce one or the other. These enterotoxins, which are both proinflamatory and cytotoxic, attack the mucosal lining of the intestines. If not identified and treated in a timely fashion, can result in permanent damage to the colon or colitis. This assay employs an antibody specific to Toxin B without cross-reactivity to toxin A.
Difficile Toxin B.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty