Would you like to log in again?
Fecal Cryptosporidium parvum Antigens ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CRY35-K01
103389-170EA 843.46 USD InStock
103389-170
Fecal Cryptosporidium parvum Antigens ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
C
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2 hours
Cryptosporidiosis is one of the main causes of persistent diarrhea in the developed world. It is caused by the presence of Cryptosporidium parvum oocysts in the gastro-intestinal tract. This parasite is known to be highly pathogenic and its infectious stage is transmitted by faecal-oral contract. It is also an opportunistic pathogen found in immunocompromised patients. The symptoms of cryptosporidiosis are watery diarrhea, stomach cramps, weight loss, nausea, and fever1. In industrialized countries, 2-2.5% of diarrhreal hospitalized patients shed C. parvum oocysts. Ten percent of AIDS patients have chronic cryptosporidiosis and this figure can be as high as 40% in certain developing countries. C. parvum is diagnosed by either Ziehl-Neelsen stain or immunofluorescence in smears of unconcentrated specimens.
parvum Antigen ELISA is intended for the qualitative detection of Cryptosporidium parvum antigen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty