Would you like to log in again?
Infliximab ELISA Assay kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IG-AA101
103389-206EA 1941.92 USD InStock
103389-206
Infliximab ELISA Assay kit, Eagle Biosciences, Inc.
Assays ELISAs
Infliximab ELISAdetermines free Infliximab in serum and plasma samples.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 5 ng/mL
  • Incubation: 2 hours
The Eagle Biosciences Infliximab ELISA Assay Kit is an enzyme immunoassay for the precise analytical determination of free Infliximabin serum and plasma samples. The Infliximab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty