Would you like to log in again?
Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IG-AA115
103389-218EA 1798.08 USD InStock
103389-218
Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Aflibercept ELISA is intended to measure free Aflibercept in serum and plasma samples.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 5 ng/mL
  • Incubation: 2 hours
The drug Aflibercept (trade names Eylea®, Zaltrap®) is a recombinant fusion protein consisting of VEGF-binding portions from the extracellular domains of human VEGF receptors 1 and 2 fused to the Fc portion of human IgG1. Aflibercept has broad affinity for all ligands that bind to these receptors, including isoforms of VEGF-A, VEGF-B, and placental growth factors. Aflibercept demonstrated antitumour effects and antiangiogenic activity as a single agent and enhanced activity in combination with chemotherapy. Identification of biomarkers for (non-)response and risk factors for adverse drug reactions that might be related to serum concentrations and maintaining the effective concentration of Aflibercept in order to potentially avoid some side effects with a reliable method might be beneficial.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty