Would you like to log in again?
Bevacizumab (mAb-based) ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IG-AB104
103389-224EA 1941.92 USD InStock
103389-224
Bevacizumab (mAb-based) ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Bevacizumab mAb-based ELISA precisely measures free Bevacizumab in serum and plasma samples.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 2 ng/mL
  • Incubation: 2 hours
The drug Bevacizumab (trade name Avastin® ) is a recombinant human IgG1::k monoclonal antibody specific for all human vascular endothelial growth factor-A (VEGF-A) isoforms and it has been approved by the FDA as a first-line treatment for metastatic colorectal cancer in combination with chemotherapy. Furthermore, VEGF is implicated in intraocular neovascularization associated with diabetic retinopathy and age-related macular degeneration. The specificity of this testsystem is achieved by using a monoclonal antibody named "MAY-2B5" for the coating of the microtiter plate. This antibody is specific for Bevacizumab only and does not cross react with other VEGF catchers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty