Would you like to log in again?
Trastuzumab ELISA Assay Kit, Herceptin®, Eagle Biosciences
Supplier: Eagle Biosciences
IG-AB105
103389-226EA 1941.92 USD InStock
103389-226
Trastuzumab ELISA Assay Kit, Herceptin®, Eagle Biosciences
Assays ELISAs
Trastuzumab (mAb-based) ELISA is for precise determination of free Trastuzumab in serum and plasma.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 2 ng/mL
  • Incubation: 2 hours
The drug Trastuzumab (trade names Herceptin® or Herclon®) is a recombinant DNA-derived humanized monoclonal antibody that selectively targets the extracellular domain of the human epidermal growth factor receptor 2 protein (HER2). The antibody is an IgG1 kappa that contains human framework regions with the complementarity-determining regions of a murine anti-p185 HER2 antibody that binds to HER2. Trastuzumab blood concentrations throughout the dosing interval expected to be remaining above those considered necessary for anticancer activity. Furthermore, in a separate analysis, patients with the lowest Trastuzumab serum trough concentrations had the highest rate of disease progression and shortest overall survival. This Eagle Biosciences ImmunoGuide Trastuzumab ELISA (mAb-based) is developed for the specific measurement of Trastuzumabin serum, plasma,and other biological fluids by the advantage of using a site-directed VME-5A6b mouse monoclonal antibody (mAb) specific for Trastuzumab only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty