Would you like to log in again?
Rituximab ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IG-AB106
103389-228EA 1941.92 USD InStock
103389-228
Rituximab ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Rituximab ELISA is a mAb-based that detects the concentration of Rituximab in serum and plasma.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 2 ng/mL
  • Incubation: 2 hours
The Eagle Biosciences Rituximab (Rituxan®, Mabthera®) ELISA Assay Kit is developed for specific measurement of Rituximab in serum and plasma.  The kit is a solid phase enzyme-linked immunosorbent assay (ELISA) is based on rituximab-specific 9F9 monoclonal antibody (mAb).  The Eagle Biosciences Rituximab (Rituxan®, Mabthera®) ELISA Assay Kit  is for research use only and is not intended for diagnostic or therapeutic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty