Would you like to log in again?
Omalizumab ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-AB111
103389-236EA 1941.92 USD InStock
103389-236
Omalizumab ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Omalizumab ELISA is for the measurement of free Omalizumab in serum and plasma samples.
  • Dynamic Range: 3 - 100 ng/mL
  • Sensitivity: 1 ng/mL
  • Incubation: 2 hours
The drug Omalizumab (trade name Xolair®) is a recombinant DNA-derived humanized IgG1κ monoclonal antibody and it binds to human immunoglobulin E (IgE). The molecular weight of Omalizumab is 149 kilodaltons and is produced by a Chinese hamster ovary cell suspension culture. Omalizumab inhibits the binding of IgE to IgE receptor (FcεRI) on the surface of mast cells and basophils. Therefore, the Omalizumab is expected to limit the degree of release of mediators of the allergic response from the FcεRI bearing cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty