Would you like to log in again?
Cetuximab (mAB-based) ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-AB112
103389-238EA 1941.92 USD InStock
103389-238
Cetuximab (mAB-based) ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Cetuximab mAb-based ELISA precisely measures free Cetuximab in serum and plasma.
  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 2 ng/ml
  • Incubation: 2 hours
The drug Cetuximab (trade name Erbitux®) is a human-mouse chimeric immunoglobulin (Ig) G1 monoclonal antibody (mAb), selectively directed against the epidermal growth factor receptor (EGFR), also known as HER1 or ErbB1. Since its approval by Food and Drug Administration for cancer treatment in 2004, Cetuximab became widely used in the treatment of colorectal and head and neck cancers. The ImmunoGuide Cetuximab ELISA  (mAb-based) kit can be efficiently used for measuring free Cetuximab levels in serum and plasma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty