Would you like to log in again?
Antibody to Etanercept ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-BB102
103389-242EA 2013.85 USD InStock
103389-242
Antibody to Etanercept ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Antibody to Etanercept ELISA to determine free antibodies to Etanercept in serum and plasma samples.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours
The Eagle Biosciences Antibody to Etanercept is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Etanercept in serum and plasma samples. The Antibody to Etanercept ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty