Would you like to log in again?
Antibody to Golimumab ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-BB107
103389-252EA 1941.92 USD InStock
103389-252
Antibody to Golimumab ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Antibody to Golimumab (Simponi) ELISA determines free antibodies to Golimumab in serum and plasma.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours
The Eagle Biosciences Antibody to Golimumab ELISA Assay Kit is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Golimumab in serum and plasma samples. The Antibody to Golimumab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty