Would you like to log in again?
Antibody to Tocilizumab ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-BB108
103389-254EA 1941.92 USD InStock
103389-254
Antibody to Tocilizumab ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Antibody to Tocilizumab ELISA determines free antibodies to Tocilizumab in serum and plasma samples.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours
The Eagle Biosciences Antibody to Tocilizumab is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Tocilizumab (Atlizumab is just another name for the same drug) in serum and plasma samples. The Antibody to Tocilizumab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty