Would you like to log in again?
Total Infliximab (Remicade®) ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-TA101
103389-260EA 1941.92 USD InStock
103389-260
Total Infliximab (Remicade®) ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Total Antibodies to Infliximab EIA detects total antibodies to Infliximab in serum & plasma samples.
  • Dynamic Range: 10 tAU/mL
  • Sensitivity: Cut-off
  • Incubation: 1.5 hours
The drug Infliximab (trade name Remicade® and Remsima®) is a chimeric monoclonal antibody and used to treat auto-immune disorders. One of the major concerns, despite of its wide usage, is the potential development of anti-Infliximab antibodies (ATI) which in turn may interfere with the drug efficacy as mainly judged by observing the relapse of signs and symptoms of disease and necessitate dose-escalation or potentially ending up the treatment.The ImmunoGuide Total Antibody to Infliximab ELISA Kit can be efficiently used for measuring free and Infliximab-bound antibodies against the drug Infliximab. In combination with the drug level determination, the Total Antibody to Infliximab ELISA Kit has been designed for the measurement of free antibodies against this drug. It also detects such antibodies which already are bound to the drug, even if the drug is present in a 20 fold excess.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty