Would you like to log in again?
Biotin (Serum/Plasma/Urine) ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
KR8141
103389-274EA 1360 USD InStock
103389-274
Biotin (Serum/Plasma/Urine) ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Biotin (Serum/Plasma/Urine) ELISA is intended to detect biotin in serum, plasma, urine and milk.
  • Dynamic Range: 48.1 - 1100 ng/L
  • Sensitivity: 32.4 ng/L
  • Incubation: 1.25 hours
Biotin (vitamin H) is present in bacterial, funghi, plants and animals. In food, the major part of biotin is covalently bound to protein, leaving only a minor fraction freely available. During digestion, biocytin (biotinyl-lysine) is released from the proteins and can be resorbed as easily as biotin from the intestinal tract. Afterwards, biotin is released from biocytin by the enzyme biocytinase in erythrocytes and plasma. It is then available as prosthetic group for a series of biotin-dependent enzymes. The daily requirements of biotin are difficult to estimate because a healthy intestinal flora supplies a major part of biotin by endogenous synthesis. The generally recommended daily dose for adults is 100–200 μg. Chronic hemodialysis patients show clear improvements of neuropathological status and glucose metabolism when supplemented with biotin in a milligram range. Biotin deficiency can be caused by e. g. destruction of the intestinal flora or extreme diets (e. g. frequent consumption of raw eggs). It can lead to dermatitis, hair loss, anorexia, mucular hypotonia, depression and reproduction problems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty