Would you like to log in again?
IGFBP-6 ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
E112
103389-280EA 699.62 USD InStock
103389-280
IGFBP-6 ELISA, Eagle Biosciences
Assays ELISAs
IGFBP-6 ELISA is for the measurement of human IGFBP-6 in human serum/plasma and other body fluids.
  • Dynamic Range: 0.1 - 10 ng/ml
  • Sensitivity: 0.026 ng/mL
  • Incubation: 3 hours
The Insulin-like Growth Factor 6 (IGFBP-6) is part of the IGF-System which consists of several binding proteins (1-6) and IGF-I and –II. The Insulin-like Growth Factors are involved in the control of human growth and regulated themselves by their binding proteins. The IGFBPs show high amino acid sequence homology, they have conserved N- and C-terminal domains which are involved in IGF-binding. In contrast the central linker region is highly variable. This region is not directly involved in IGF-binding but influences stability and localisation of the IGFBP / IGF complex and can be modified by glycosylation and phosphorylation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty