Would you like to log in again?
SeroELISA Chlamydia IgG ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
111-01
103389-314EA 647.31 USD InStock
103389-314
SeroELISA Chlamydia IgG ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
This SeroELISA IgG is intended to determine specific IgG antibody to Chlamydia.
  • Dynamic Range: 0.02 - 2.0 ng/mL
  • Sensitivity: 0.02 ng/mL
  • Incubation: 2 hours
Chlamydia is a gram-negative obligate intracellular bacteria that causes acute and chronic diseases in mammalian and avian species. The genus Chlamydia is comprised of four species: C.trachomatis, C.pneumoniae, C.psittaci and C. pecorum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty