Would you like to log in again?
SeroCT C. trachomatis IgG ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
181-01
103389-334EA 588.46 USD InStock
103389-334
SeroCT C. trachomatis IgG ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
SeroCT IgG ELISA is intended to detect IgG antibodies specific to C
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.25 hours
Chlamydia is a gram negative obligate intracellular bacteria that causes acute and chronic disease in mammalian and avian species. The genus Chlamydia is comprised of four species: C. trachomatis, C. pneumoniae,C. psittaci and C. pecorum. C. trachomatis is divided into 15 serovars. Serovars A, B, Ba and C are agents of trachoma(9), the leading cause of preventable blindness, endemic in third world countries. Serovars L1-L3 are the agents of lymphogranuloma venereum. Serovars D-K are the common cause of sexually transmitted genital infection worldwide: cervicitis, endometritis/salpingitis in females and urethritis(11) in both males and females. Endometritis/salpingitis can lead to tubal occlusion with a higher risk of extrauterine pregnancy and infertility. Genital infection may cause an acute and persistent infection occasionally without any clinical signs. Generally, these infections are treatable once they are diagnosed. However, without any treatment the infection may progress to a severe chronic inflammation leading to infertility, ectopic pregnancy, induced abortion or child delivery. Moreover, infants to infected mothers can be infected during birth leading to conjunctivitis or pneumonia. The serology of C. trachomatis is more interesting in cases of chronic infections than in acute infections.
trachomatis in human serum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty