Would you like to log in again?
Rat Glicentin ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
YK111
103389-362EA 1386.15 USD InStock
103389-362
Rat Glicentin ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Rat Glicentin ELISA is to determine the concentration of rat glicentin in plasma samples.
  • Dynamic Range: 2.06 - 50 pmol/mL
  • Sensitivity: 2.06 pmol/mL
  • Incubation: 20 hours
Glicentin is a 69-amino-acid peptide containing glucagon and oxyntomodulin sequences in the molecule. It is suggested that glicentin and oxyntomodulin are produced in the intestinal L-cells and glucagon in A-cells in the pancreas, these peptides are derived from a common precursor by two different tissue-specific processing pathways. In 1983, the amino acid sequence of human glicentin was deduced by Bell et al. from the genomic sequence of human preproglucagon. Glicentin is a major form of gut glucagon-like immunoreactants (Gut GLIs). In mammalian small intestine, proglucagon is processed into glicentin, oxyntomodulin, and glucagon –like peptide (GLP)-1 and –2. GLP-1(7-37) and GLP-1(7-36) amide have been isolated from the intestine and pancreas. It has been known that the GLP-1 sequence is well conserved between species in all mammals studied. Using synthetic peptides, several investigators have demonstrated that in contrast to GLP-1(1-37), truncated GLP-1(7-36) amide and GLP-1(7-37) have several physiological effects. However, the physiological role of glicentin, a major gut glucagon, is still unclear. It has been known that the circulating level of plasma glicentin-like peptides increases significantly nutrient ingestion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty