Would you like to log in again?
RAT GLP-2 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
YK140
103389-364EA 1464.62 USD InStock
103389-364
RAT GLP-2 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Rat GLP-2 ELISA is to determine the concentration of rat GLP-2 in serum or plasma samples.
  • Dynamic Range: 0.137 - 100 ng/mL
  • Sensitivity: 0.137 ng/mL
  • Incubation: 20 hours
Tissue-specific posttranslational processing of proglucagon by the prohormone convertase produced the different proglucagon derived peptides (PGDPs) in both pancreas and intestine. The most notable pancreatic PGDP is glucagon, whereas the L cell produces several structurally related peptides, including glucagon-like peptide 1 (GLP-1) and GLP-2, as well as glicentin and oxyntomodulin, which contain glucagon sequence in their molecules. Among PGDPs, GLP-2 has recently been found to show intestinal epithelial proliferation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty