Would you like to log in again?
Human VEGF ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
VGF31-K01
103389-376EA 660.38 USD InStock
103389-376
Human VEGF ELISA KIT, Eagle Biosciences
Assays ELISAs
VEGF ELISA detects human Vascular Endothelial Growth Factor concentrations in biological samples.
  • Dynamic Range: 31.25 - 1000 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 3.5 hours
The Eagle Biosciences Human Vascular Endothelial Growth Factor (VEGF) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Vascular Endothelial Growth Factor (VEGF) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Vascular Endothelial Growth Factor (VEGF) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty