Would you like to log in again?
Human MMP-9 ELISA KIT, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
MM931-K01
103389-378EA 660.38 USD InStock
103389-378
Human MMP-9 ELISA KIT, Eagle Biosciences, Inc.
Assays ELISAs
MMP-9 ELISA detects human Matrix Metalloproteinase 9 concentrations in cell culture,serum, & plasma.
  • Dynamic Range: 62.5 - 2000 pg/ml
  • Sensitivity: 31 pg/ml
  • Incubation: 3.5 hours
The Eagle Biosciences Human Matrix Metalloproteinase 9 (MMP-9) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Matrix Metalloproteinase 9 (MMP-9) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Matrix Metalloproteinase 9 (MMP-9) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty