Would you like to log in again?
DKK-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20413
103389-386EA 954.62 USD InStock
103389-386
DKK-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human DKK-1 ELISA is to detect Human DKK-1 in serum.
  • Dynamic Range: 1.56-50 ng/ml
  • Sensitivity: 0.6 ng/ml
  • Incubation: 1h40
Dickkopf-1(DKK-1) is a 28.67 kDa secreted protein that acts as soluble inhibitor of the WNT signalling pathway. This pathway contains lipid-modified glycoproteins that activate cell surface receptor-mediated signal transduction to regulate cell activities like: cell fate, proliferation, migration, polarity and gene expression. DKK-1 regulates developmental processes of all kinds. Thus, DKK-1 is also involved in the regulation of bone metabolism as it effects osteoblast differentiation and in regulation of tumourgenetic activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty