Would you like to log in again?
NT-proBNP ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
SK-1204
103389-396EA 993.85 USD InStock
103389-396
NT-proBNP ELISA, Eagle Biosciences
Assays ELISAs
Human NT-proBNP ELISAdetermines human NT-proBNP concentrations in serum.
  • Dynamic Range: 0- 640 pmol/L
  • Sensitivity: 3 pmol/L
  • Incubation: 3.5 hours
The Eagle Biosciences NT-proBNP ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Nt-proBNP in serum. The NT-proBNP ELISA Assay Kit is manufactured by Biomedica.  The NT-proBNP ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty