Would you like to log in again?
Universal Protease Activity ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
30516103
103389-404EA 1477.69 USD InStock
103389-404
Universal Protease Activity ELISA, Eagle Biosciences
Assays ELISAs
Universal Protease Activity ELISA measures activity of proteases like MMPs, ADAMTS, and HtrA.
  • Dynamic Range: 0.312 - 20 nM
  • Sensitivity: 312 pM HtrA1
  • Incubation: 2.25 hours
Proteases play important roles in the control of multiple biological processes in all living organisms. They regulate the activities of many proteins, modulate protein-protein interactions, create new bioactive molecules, contribute to the processing of cellular information and generate, transduce and amplify molecular signals. As a direct result of these multiple actions, proteases influence DNA replication and transcription, cell proliferation and differentiation, tissue morphogenesis and remodelling, heat shock and unfolded protein responses, angiogenesis, neurogenesis, ovulation, fertilization, wound repair, stem cell mobilization, haemostasis, blood coagulation, inflammation, immunity, autophagy, senescence, necrosis and apoptosis. Consistent with these essential roles of proteases in cell behaviour, survival and death of all organisms, alterations in proteolytic systems underlie multiple pathological conditions such as cancer, neurodegenerative disorders, inflammatory and cardiovascular diseases. The Universal Protease Activity ELISA is based on a universal substrate for proteases, beta casein which is labeled with biotin and coupled on beads. The cleaved fragments of biotinylated beta casein are detected with an enzyme linked immuno assay (ELISA). The combinations of bead technology and ELISA have a synergistic effect. The sensitivity and specificity via antibodies is a feature of ELISA and the beads have no scale limit and can be used in variable volumes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty