Would you like to log in again?
MMP-13 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
30518101
103389-410EA 1144.23 USD InStock
103389-410
MMP-13 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
MMP-13 ELISA is sensitive and specific to determine activated human MMP-13 in biological samples.
  • Dynamic Range: 32 -2000 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 4.25 hours
Matrix Metalloproteinases (MMPs) comprise a family of secreted and membrane-bound endopeptidases which hydrolyse extracellular matrix proteins (1). Based on their preferred substrates and on structural features MMPs can be divided into collagenases, gelatinases, stromelysins and membrane-type matrix metalloproteinases. Their activity is tightly regulated by inhibitors (2). Collagenase 3 (MMP-13) is a secreted 452 residue protein which is released from cells as an inactive zymogen and activated extracellularly by removal of its propeptide. Cleavage of the 84 residue propeptide can be catalysed by other MMPs such as MMP-2 and MMP-14, or by other proteases like plasmin (3). Under normal conditions, MMP-13 is expressed during embryogenesis (fetal bone development) and is present only at very low levels in adult tissue. However, this enzyme is reported to be involved in the development and metastasis of breast and lung carcinomas, chondrosarcomas and osteosarcomas, head and neck carcinomas and some forms of skin cancer (4). Additionally, this enzyme plays an important role in degenerative bone diseases including osteoarthritis (5) and rheumatoid arthritis (6). Our MMP-13 test kit enables the user to measure the concentration of this putative marker enzyme in body fluids such as serum and synovial fluid with high sensitivity. The ELISA MMP-13 is specific for the activated enzyme.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty