Would you like to log in again?
Electrode Cleaning Solution, pH/ORP, Oakton Instruments
Supplier: Cole Parmer
00653-06
103391-788EA 18.79 USD InStock
103391-788
Electrode Cleaning Solution, pH/ORP, Oakton Instruments
Electrolytes Electrode Storage Solutions
General-purpose pH/ORP electrode cleaning solution removes build-up from electrodes and maintains electrode sensitivity.
  • Extends the life of electrodes
  • Maintain accurate readings
  • Use the reference fill solutions with refillable pH electrodes
The solution accepts pH electrodes up to 12 mm in Ø.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty