Would you like to log in again?
SHuffle T7 Express Competent E. coli, New England Biolabs
Supplier: New England Biolabs
C3029J
103391-806EA 264 USD InStock
103391-806
SHuffle T7 Express Competent E. coli, New England Biolabs
Competent Cells
Chemically competent E. coli cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm.
  • T7 expression
  • Engineered E. coli B strain to promote disulfide bond formation in the cytoplasm
  • Deficient in proteases Lon and OmpT
  • Resistant to phage T1 (fhuA2)
  • Free of animal products
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty