Would you like to log in again?
Antibody to Vedolizumab ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-BB116
103491-760EA 1941.92 USD InStock
103491-760
Antibody to Vedolizumab ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Vedolizumab Entyvio Antibodies ELISA determines free antibodies to Vedolizumab in serum and plasma.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 1.5 hours
The drug Vedolizumab (trade name Entyvio®) is a humanised immunoglobulin G1 monoclonal antibody that binds exclusively to the lymphocyte integrin α4β7, is indicated for the treatment of moderately to severely active ulcerative colitis [UC] or Crohn’s disease [CD]. During treatment the presence of persistent anti-Vedolizumab antibody was observed to substantially reduce serum concentrations of Vedolizumab, either to undetectable or negligible levels. The Eagle Biosciences Antibody to Vedolizumab ELISA Assay kit has been designed for the measurement of free antibodies against this drug. It does not detect such antibodies which already are bound to the drug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty