Would you like to log in again?
BioProfile® Flex 2 Microsensor Cards for Automated Chemistry Analyzers, Nova Biomedical
Supplier: Nova Biomedical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
57528 42458 57510 57526 57512
103497-222EA 646.54 USD InStock
103497-222 103497-220 103497-226 101447-466 103497-224
BioProfile® Flex 2 Microsensor Cards for Automated Chemistry Analyzers, Nova Biomedical
Electrodes Other Electrodes
BioProfile® Flex 2 Microsensor cards are mainly used for automated chemistry analyzers. Chemistry and gas biosensors are combined in the credit card sized MicroSensor Card, which uses biosensor technology that has been validated in thousands of cell culture processes.
  • Maintenance-free
  • Replaceable in seconds
pH/Gas microsensor cards sense the parameters such as pH (8.50 to 12), pCO2 (25 to 8) and pO2 (4 to 10).
Chemistry microsensor cards sense the parameters such as Glucose (2 to 100), NH4 (8.2 to 12.5), Na (8.2 to 12), K (8.2 to 12) and Ca (8.2 to 12).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty