Would you like to log in again?
QuickSwitch™ Quant H-2 Kb Tetramer Kit-APC, MBL International
Supplier: MBL International
TB-7400-K2
103501-248EA 2307.69 USD InStock
103501-248
QuickSwitch™ Quant H-2 Kb Tetramer Kit-APC, MBL International
Assays Immunological Assays
QuickSwitch™ Tetramer Kit utilizes a patented technique for exchanging up to ten peptides on an MHC class I tetramer. Reduce time and cost with this kit to create your own custom tetramers. The resulting custom tetramers can be used for multiple applications, including screening immunogenic peptides.
  • Kit Components:
  • Tetramer with an irrelevant exchangeable peptide in the MHC groove
  • A peptide exchange factor for catalyzing the peptide exchange reaction
  • A high affinity MHC-binding reference peptide used as peptide exchange positive control FITC conjugated antibody specific of the exiting peptide
  • Magnetic beads conjugated with anti MHC antibody for tetramer capture
  • An Assay Buffer for diluting reagents and washing steps
Research Area: Immunology, Immune Monitoring
Quantification Included
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty