Would you like to log in again?
KLD
Supplier: New England Biolabs
M0554S
103501-322EA 354 USD InStock
103501-322
KLD
Enzymes
KLD Enzyme Mix is a unique blend of Kinase, Ligase and DpnI enzymes.
  • Degradation of template DNA by DpnI
  • Also available as a kit: with and without competent cells
This formulation allows efficient phosphorylation, intramolecular ligation/circularization and template removal in a single 5 minute reaction step at room temperature.
This master mix is a component of the Q5 Site-Directed Mutagenesis Kits and it has been designed for use with fragments that have been PCR-amplified by Q5 Hot Start High-Fidelity DNA Polymerase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty