Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNext rRNA Depletion Kit (Human/Mouse/Rat) with RNA Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs
E6350S E6350L E6350X
103501-482EA 390 USD InStock
103501-482 103501-480 103501-484
NEBNext rRNA Depletion Kit (Human/Mouse/Rat) with RNA Sample Purification Beads, New England Biolabs
Assays Nucleic Acid Assays
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) with RNA Sample Purification Beads employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human, mouse and rat total RNA preparations.
  • Compatible with a broad range of input amounts: 10 ng to 1 µg
  • Suitable for low-quality or high-quality RNA
  • Includes Agencourt RNAClean XP beads
This product is suitable for both intact and degraded RNA (e.g., FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications. For added convenience, this kit contains NEBNext RNA Sample Purification Beads (Agencourt RNAClean XP beads from Beckman Coulter).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty