Would you like to log in again?
PathHunter® eXpress DRD3 U2OS β-Arrestin GPCR Assay, DiscoverX
Supplier: Eurofins DiscoverX Corporation
93-0591E3CP0M
103502-106KT 2614.69 USD InStock
103502-106
PathHunter® eXpress DRD3 U2OS β-Arrestin GPCR Assay, DiscoverX
Assays Cellular Assays
Access DiscoverX's extensive menu of functional, whole cell GPCR, Kinase, Interleukin, Pathway, NHR, and more assays without investing in a clonal cell line
  • Complete off-the-shelf assay solution; Pre-validated frozen cells and reagents; Fast and simple step-by-step protocol; and Compatible with any standard luminometer
Please refer to the product-specific datasheet for the appropriate storage conditions for these products.
Our ultra convenient eXpress™ kits are ready to use and contain all the necessary reagents to perform functional whole cell assays - in just a few hours. Kits include frozen, pre-validated cells, detection reagents, seeding media and plates and do not require expensive instrumentation to run. Minimize assay development time and eliminate lengthy, expensive and time consuming cell culture. Simply thaw, plate and run the assay!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty