Would you like to log in again?
NEBNext® Ultra™ II FS DNA Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs
NEBNext® Ultra™
E7805L E7805S
103529-168EA 2521 USD InStock
103529-168 103529-140
NEBNext® Ultra™ II FS DNA Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext® Ultra™ II FS DNA Library Prep Kit for Illumina provides a fragmentation system that is a fast and reliable solution for DNA fragmentation and library construction.
  • Perform fragmentation, end repair and dA-tailing with a single enzyme mix
  • Experience reliable fragmentation with a single protocol, regardless of DNA input amount or GC content
  • Generate high yields with increased reaction efficiencies and minimized sample loss
  • Use with DNA in standard buffers (TE, Tris-HCl) and water
  • Save time with a streamlined workflow: ~2.5 hours, with <15 minutes hands-on time
  • Vary incubation time to generate a wide range of insert sizes
A new DNA fragmentation reagent is combined with end repair and dA-tailing reagents, allowing these steps to be performed in the same tube, with no clean-ups or sample loss. The same fragmentation protocol is used for any input amount (100 to 500 ng), or GC content.
Delivery information: kit includes the NEBNext Ultra II Ligation Master Mix for adaptor ligation, the NEBNext Ultra II Q5® Master Mix for uniform, high-fidelity library amplification, and the gold standard SPRIselect size selection and clean-up beads.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty