Would you like to log in again?
Human Recombinant Coag Factor Xiii A Chain (C-6His)
Supplier: Bon Opus Biosciences
C880-1MG C880-500UG C880-10UG C880-50UG
103529-526EA 2035.38 USD InStock
103529-526 103529-434 103529-300 103529-224
Human Recombinant Coag Factor Xiii A Chain (C-6His)
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Coagulation Factor XIII A Chain is produced by our Mammalian expression system and the target gene encoding Gly39-Met732 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty