Would you like to log in again?
Streptococcus pyogenes Cas9 Nuclease, OZ Biosciences
Supplier: OZ Biosciences
CAS9050 CAS9100 CAS9500
103537-320EA 1046.4 USD InStock
103537-320 103537-352 103537-318
Streptococcus pyogenes Cas9 Nuclease, OZ Biosciences
Enzymes
Optimized Cas9 Nuclease S. Pyogenes is designed for genome editing in living cells or organisms and also for in vitro digestion.
  • Increased genome editing efficiency using Cas9/RNP delivery
  • Successful CRISPR/Cas9 genome editing can be performed through diverse approaches (plasmids, mRNA, nuclease, viral delivery)
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). These features make nuclease protein delivery particularly well suited for precision genome engineering.
Efficient nucleic acid delivery represents a critical step for genome editing experiments.
Delivery information: Shipped with dry ice.
Caution: Cas9 nuclease and Cas9 reaction buffer must be stored at –20 °C. Avoid freeze/thawing cycles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty