Would you like to log in again?
ViroMag™ Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
ViroMag™
KC30600 KC30500 VM41000 VM40100 VM40200
103549-566EA 954 USD InStock
103549-566 103258-252 103258-250 103258-136 103258-404
ViroMag™ Transduction Reagent, OZ Biosciences
Transfection Reagents
ViroMag Transduction Reagent is a magnetic nanoparticles formulation dedicated to increase virus infection and transduction capacities; it is suitable for all viruses.
  • Increases transduction efficiency
  • Accelerates the transduction process
  • Efficient for hard-to-infect and non-permissive cells
  • Concentrates the viral dose onto the cells
  • Suitable for all type of virus
  • Synchronise cells adsorption/infection without modifications of the virus
Based on the Magnetofection™ technology, this reagent allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus transduction with extremely low vector doses. Due to its specific properties, ViroMag is ideal to infect non permissive cells.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty