Would you like to log in again?
Magnetofection Starting Kit, OZ Biosciences
Supplier: OZ Biosciences
KC30200
103549-572EA 928.8 USD InStock
103549-572
Magnetofection Starting Kit, OZ Biosciences
Transfection Reagents
Magnetofection is a simple and highly efficient transfection method to transfect primary cells and hard to transfect cells.
  • Highly efficient for primary cells
  • Powerful for hard to transfect cells
  • Non-toxic
  • Serum-compatible
  • Simple, rapid, and easy-to-use
  • For all types of nucleic acids
Inspired by the validated and recognized magnetic drug targeting technology, this original method is a revolution for transfection and infection. In essence, the idea was to unite the advantages of the popular biochemical (cationic lipids or polymers) and physical (electroporation, gene gun) transfection methods in one system while excluding their inconveniences (low efficiency, toxicity, difficulty to handle). It is the unique technology suitable for viral and non viral gene delivery applications.
Delivery information: Shipped at room temperature.
Packaging: Store at +4°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty