Would you like to log in again?
HYPE-CHO Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
HYPE-CHO™
HYC01500 HYC15000 HYC03000 HYC30000
103549-578EA 2109.02 USD InStock
103549-578 103549-574 103549-580
HYPE-CHO Transfection Kit, OZ Biosciences
Transfection Reagents
HYPE-CHO™ Transfection Kit has been designed for large scale up transient transfection and high protein expression such as antibody. The system is optimized for maximum efficiency in all CHO-S cells.
  • Reach optimal Protein & Antibody production yield
  • Suitable for all CHO cells growing in suspension
  • Compatible for any synthetic or regular media used for protein production
  • Ideal for bioreactor, spinner or flasks
  • Animal origin free
It has been used and validated with cells from different origins (CHO, CHO-S, rCHO or any CHO-related cells) cultured in suspension (flask, spinner and bioreactor) used to produce proteins.
Delivery information: Shipped at room temperature.
Packaging: Store Hype-CHO Reagent +4 °C - BCHO +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty