Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DMNPE-caged Luciferin

Supplier: OZ Biosciences
Synonyms: D-Luciferin 1-(4,5-dimethoxy-2-nitrophenyl)ethyl ester

Packaged under nitrogen.

LC10000 LC50000 LC25000
103549-586EA 420.4 USD InStock
103549-586 103549-584 103549-590
DMNPE-caged Luciferin
DMNPE-caged Luciferin
The cell permeable DMNPE-caged Luciferin is as a bioluminescent luciferase substrate used to measure intracellular functions since it readily crosses cell membranes. It is a D-luciferin ester analogue of luciferin.

  • High purity
  • Cell permeable: efficient luciferin delivery into intact cells even at neutral pH
  • Ideal for in vivo applications
  • Induces prolonged release of luciferin by the action of cells’ esterases
  • Allows to follow changes in gene expression in live cells
  • Suitable for in vitro experiments

Once the caged luciferin is inside the cells, active luciferin can be released by the action of endogenous intracellular esterases or by a flash of UV light. This implies:
1. A more efficient delivery of luciferin into intact living cells & improved sensitivity analysis of in vivo luciferase assays
2. A long-term measurement of luciferase activity since hydrolysis by esterases allows a continuous supply of active Luciferin
DMNPE-caged luciferin appears a good alternative to D-Luciferin classically used to measure luciferase activity in live cells. Indeed, the difficulty to deliver D-Luciferin into living cells renders in vivo quantification limited to a certain extent- for example, permeability issues at neurtral pH.

Shipped at room temperature.
Store at -20°C and protect from light.


Formula: C₂₁H₁₉N₃O₇S₂
MW: 489.52 g/mol
Storage Temperature: Freezer
CAS Number: 223920-67-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/88/23463188.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty