Would you like to log in again?
Thin Walled 'L' Series NMR Sample Tubes, 5 mm, New Era™
Supplier: New Era
NEUL58 NEHL58 NEHL57 NESL57 NEML57 NELL57 NEUL57 NELL58 NEML58
103549-644EA 17.83 USD InStock
103549-644 100165-940 100165-972 100165-982 100165-988 100165-942 100165-956 100165-934 100165-978
Thin Walled 'L' Series NMR Sample Tubes, 5 mm, New Era™
Tubes NMR Tubes
Tubes are specifically designed for QC, routine analysis and research applications.
  • Made of type 1, class B borosilicate glass
  • Round bottom design
  • Tubes are not suitable for flame sealing to Pyrex® or equivalent
  • Each tube is capped and marked with the catalog number
Ideal for biological samples, hands-on and high throughput/autosampler use and samples not requiring special handling. Suitable for all experiments when matched to the application and instrument field strength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty