Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Hydrogen Generator, Precision Hydrogen Trace 250 GC Carrier Gas, Peak Scientific
Supplier: Peak Scientific
64-0250
103549-776EA 17808.42 USD InStock
103549-776
Hydrogen Generator, Precision Hydrogen Trace 250 GC Carrier Gas, Peak Scientific
Gas Generators Hydrogen Generators
The Precision Hydrogen Trace 250 generator is designed primarily for GC carrier gas use, and can also be used for detectors, requiring a hydrogen on demand solution for fuel gas such as FID and FPD. One generator is capable of supplying multiple GC instruments.
  • Suitable for flame gas and carrier gas at trace detection limits
  • 99.99999%* purity
  • Internal leak detection with automatic shutdown features
  • Proven PEM technology to generate hydrogen safely and reliably
  • Regenerative PSA dryers to ensure highest level of purity
  • Automatic loading pump as standard
  • Maintenance limited to replacing de-ionizer cartridge
  • Compact, space-saving modular design
  • Creates hydrogen on demand, minimal storage of hydrogen in the system
  • Combine multiple units for higher flow requirements
  • GC in-oven hydrogen leak detector available as an optional extra
  • Peak offers a 3 year cell warranty with this generator as standard
The Precision Hydrogen Trace 250 generator is designed primarily for GC carrier gas use, and can also be used for detectors, requiring a hydrogen on demand solution for fuel gas such as FID and FPD. One generator is capable of supplying multiple GC instruments..
This generator offers a safe, reliable and convenient solution for those using hydrogen for GC carrier gas, producing the highest purity of hydrogen from the Precision Series by utilising a Proton Exchange Membrane to create the hydrogen gas from deionized water, as well as Pressure Swing Absorption and molecular sieve technology to remove moisture content down to trace levels. The Precision Hydrogen Trace generators are also suitable for providing collision gas for ICP-MS.
The Precision Hydrogen Trace 250 generator also comes with many highly robust safety features as standard, allowing for peace of mind in the laboratory and a far safer, dependable and more convenient alternative to cylinder gas
Caution: Based on O₂ content independently verified by National Physical Laboratory, UK
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty