Would you like to log in again?
Helix-IN, Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Helix-IN™
HX10100 HX11000 HX10500
103565-924EA 270.35 USD InStock
103565-924 103565-922 103565-926
Helix-IN, Transfection Reagent, OZ Biosciences
Transfection Reagents
OZ Biosciences revolutionizes Polyfection with the design of Helix-IN™, a novel patented Cationic Hydroxylated Amphiphilic Multi-block Polymer (CHAMP™ Technology). This novel bi-functional co-polymer is biocompatible, ionizable, pH responsive and biodegradable.
  • Broad Spectrum Transfection Reagent for cell lines and hard-to-transfect cells
  • High transfection efficiency & increased transgene expression even with low DNA amount
  • High intracellular protein production while preserving viability
  • High secreted protein production while minimizing cellular stress
  • Biodegradable-Avoid secondary effects
  • Compatible with any culture medium
Delivery information: Shipped at room temperature.
Packaging: Store at -20°C
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty