Would you like to log in again?
Biacore Maintenance Kit (Type 3), Cytiva
Supplier: Cytiva
29229054
103568-398EA 849.15 USD InStock
103568-398
Biacore Maintenance Kit (Type 3), Cytiva
Buffers Biological Buffers
Kit for conducting instrument maintenance of Biacore 8K SPR system, containing BIAtest, BIAdesorb and BIAnormalizing solutions.
BIAnormalizing solution (70% (w/w) glycerol is sufficient for 2 months maintenance for Biacore 8K of normal use together with the other components. Used with all Biacore instruments, except Biacore C.
Due to the Biocidal products regulation, the BIAdisinfectant solution (containing sodium hypochlorite) is not part of Biacore maintenance kit, type 3.
Delivery information: Kit contains: BIAtest solution (HBS-EP) 65 ml, BIAnormalizing solution (70%) 90 ml, 1×Series S maintenance chip, 2×BIAdesorb solution 1500 ml and 2×BIAdesorb solution 2500 ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty